Test strips, Glucose, QUANTOFIX®
Fournisseur: MACHEREY-NAGEL
91348
MANA91348EA 33.2 EUR InStock
MANA91348
Test strips, Glucose, QUANTOFIX®
Kits de tests environnementaux
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips allow the quick and easy determination of glucose in solutions. The easy dip-and-read-procedure provides reliable results in less than 30 seconds. There is absolutely no training required.
Glucose is an important part of many foods and beverages and hence it's determination is important in the respective industries. The test is specially suited for the potato industry, to provide quick and easy quality control procedures at the initial deliveries. Additionally, the glucose content in potatoes is an important quality criterion during storage, as the glucose content increases during long storage or at high temperatures, leading to loss of quality, discolorations and unpleasent taste. These test strips are the ideal solution to test for glucose in solution or vegetables quickly and accurately. The test can also be run on the QUANTOFIX® Relax.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité