Test strips, Active Oxygen, QUANTOFIX®
Fournisseur: MACHEREY-NAGEL
91349
MANA91349EA 37.5 EUR InStock
MANA91349
Test strips, Active Oxygen, QUANTOFIX®
Kits de tests environnementaux
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips are for the fast and easy determination of active oxygen or potassium monopersulfate (KMPS) respectively. The easy dip-and-read-procedure provides reliable results within 30 seconds. So-called active oxygen is used in swimming pools as an alternative to chlorine for disinfection purposes. Therefore KMPS is often added to the water. The respective content can be easily determined with the test strips to assure a safe disinfection.
Gradation: 0 · 4 · 8 · 15 · 25 mg/L KMPS
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité