Test strips, Silver, QUANTOFIX®
Fournisseur: MACHEREY-NAGEL
91350
MANA91350EA 27.6 EUR InStock
MANA91350
Test strips, Silver, QUANTOFIX®
Kits de tests environnementaux
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips allow the quick and easy determination of silver in solutions. The easy dip-and-read-procedure provides reliable results in less than 15 seconds. There is absolutely no training required.
Silver is an universally known and important metal, which is used in many applications. Silver plays an important role in the final stages of photo and film development, the so-called fixing process. Here, excess silver halides are washed from the photographic layer. To ensure a well functioning fixing bath, the silver content needs to be monitored regularly. These test strips are the ideal tool to test for silver in solutions quickly and accurately.
Gradation: 0 · 1 · 2 · 3 · 5 · 7 · 10 g/l Ag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité