Titrimetric rapid tests, acidity, VISOCOLOR®
Fournisseur: MACHEREY-NAGEL
915006
MANA915006EA 54.4 EUR InStock
MANA915006
Titrimetric rapid tests, acidity, VISOCOLOR®
Kits de tests environnementaux
The VISOCOLOR® testkits are devided into VISOCOLOR® alpha, ECO and HE. Every product range offering its specific strenght to provide a tailored test for each and every user and application.
  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges
In natural unpolluted waters mainly carbonic acid is present, but also humic acids. With this test kit all acids, i. e. also those present in industrial waters, can be determined.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité