Reagent set (7 parameters), VISOCOLOR® ECO
Fournisseur: MACHEREY-NAGEL
931301
MANA931301EA 271 EUR InStock
MANA931301
Reagent set (7 parameters), VISOCOLOR® ECO
Kits de tests environnementaux
Reagent cases are flexible tools for all areas of water and soil analysis. The cases are ideal for testing water bodies, for fish farming and a wide variety of other applications  within the area of environmental  management and water monitoring.
  • Rugged cases with premium foam inlays
  • Pre-packed or empty cases for individual solutions
The reagent cases enable users to get results fast and directly at the point of interest. Previous chemical knowledge or experience is not required to run any of the tests or to use the cases effectively. 
Information de livraison: Kit contains ammonium, carbonate hardness, total hardness, nitrate, nitrite, pH, phosphate 9 test kits and  include all necessary test instructions and analytical accessories for especially easy and convenient handling.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité