Reagent case for soil analysis, VISOCOLOR®
Fournisseur: MACHEREY-NAGEL
934220
MANA934220EA 1225 EUR InStock
MANA934220
Reagent case for soil analysis, VISOCOLOR®
Kits de tests environnementaux
This portable laboratory case contains all reagents, instruments and accessories required for the preparation of soil extracts and the subsequent analysis of phosphate (P), potassium (K), ammonium, nitrate, nitrite (N), soil structure and pH.
  • Designed for rapid, convenient and reliable soil analysis in the lab or in the field
  • Measurements are carried out either with colorimetric rapid tests or with easy to use test strips
Informations de commande: NANOCOLOR® reagents and photometers must be ordered separately.
Commander maintenant
COMMANDE
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité