Test kits, residual hardness, VISOCOLOR® alpha
Fournisseur: MACHEREY-NAGEL
935080
MANA935080EA 44.1 EUR InStock
MANA935080
Test kits, residual hardness, VISOCOLOR® alpha
Kits de tests environnementaux
This kit is designed for the colorimetric determination of residual hardness.
The total hardness of water is based on its content of alkaline earth ions (calcium and magnesium ions). This content depends on the geological conditions the water encounters during its course and may vary widely. Knowledge of the total hardness is important for the use of water in industrial as well as municipal applications, e.g. in the household as wash water or as boiler feed water in industry.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité