NANOCOLOR® ECO Photometric Reagent Kit, Iron
Fournisseur: MACHEREY-NAGEL
976002
MANA976002EA 132 EUR InStock
MANA976002
NANOCOLOR® ECO Photometric Reagent Kit, Iron
Kits de tests environnementaux
Reagent kit for the determination of iron. Precise rapid tests for all kind of water and waste water samples. Time saving and reliable analysis together with our NANOCOLOR photometers.
  • Sea water analysis
  • Shelf life for 1,5 year(s)
  • Reagents for 100 tests in a box
Iron is ubiquitous in our daily lives. Iron salts are essential for human life and are absorbed through food and drinking water. However, excessive concentrations of iron in water are undesirable due to discoloration and the formation of deposits such as rust.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité