SCRAPEX 596 Scraper
Fournisseur: MARTOR ARGENTAX
596.03
MARO596.03EA 33.4 EUR InStock
MARO596.03
SCRAPEX 596 Scraper
Cutters
The SCRAPEX 596 is the largest scraper used for effortless removal of stickers, labels and other paper, colour and paint residue on all smooth surfaces. It can work especially effectively with the 100 mm long blade. This professional tool also convinces with a non-slip handle and the built-in safeguard.
  • Robust and durable
  • Broad user surface
  • Safeguard included
  • Non-slip handle
  • Easy blade changing
The 11 cm long handle is made of die-cast zinc, which makes it robust and durable. The handle is equipped with a non-slip soft grip and small pores that are friendly to the hands. And the special screw will also help you change blades easily.
Conditionnement: One in self-service single unit box.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité