Thermo Scientific™ Capit-All™ Flex Automated Decapper
Fournisseur: THERMO MATRIX TECHNOLOGIES
4120-FLEX
MATR4120-FLEXEA 81380 EUR InStock
MATR4120-FLEX
Thermo Scientific™ Capit-All™ Flex Automated Decapper
Pinces à sertir et pinces à dessertir
A full-service, automated capping and decapping solution for Thermo Scientific™ Matrix™ and Nunc™ 48 and 96 tube racks.
  • Flexibility: Designed for use as a standalone benchtop unit or in an integrated system
  • Compatible with Matrix and Nunc tubes in 48- and 96-tube formats, can switch between tube formats without adapters, supporting both current and changing application requirements
  • Efficiency: Completes a capping cycle in less than 60 seconds, can handle both full and partial racks of tubes, offering flexibility for varying workflows
  • Angled, 7" touchscreen with user-friendly interface, enables easy program selection with just a few clicks, even with gloves
  • Exceptional sample protection: Includes a drip tray that acts as a barrier between caps and tubes to help prevent cross-contamination and maintain sample integrity
The Capit-All Flex automated decapper delivers the flexibility needed to power genomics, biobanking, precision medicine, and other storage applications while helping reduce your laboratory’s physical footprint and increasing efficiency.
Informations de commande: For a selection of compatible tubes, see the Storage tubes, 2D barcoded, Matrix™, Storage tubes, low volume, 2D barcoded, Matrix™, Matrix™ Tri-Coded Tubes, Cryobank vials and Bank-It™ tube system, Nunc™, and Nunc™ Internally-Threaded Universal Tubes and Rack System series.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité