Foetal bovine serum, Australian origin, Corning®
Fournisseur: Corning
35-076-CV
MDTC35-076-CVEA 992.51 EUR InStock
MDTC35-076-CV
Foetal bovine serum, Australian origin, Corning®
Sérums
Foetal bovine serum sourced in Australia and manufactured at facilities approved by the appropriate local authorities.
  • Triple sterile filtered (0,1 μm)
  • Low endotoxin (<20 EU/ml)
  • Low haemoglobin (<30 mg/dl)
Corning® sera products are analysed for quality and performance. Testing includes pH, osmolality, growth promotion, sterility (current USP method), mycoplasma, total protein, endotoxin and haemogloblin levels, and viral contamination. A full biochemical profile is available for each lot upon request.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité