Protein and serum free medium, PF medium, Corning®
Fournisseur: Corning
40-200-CV
MDTC40-200-CVEA 192.94 EUR InStock
MDTC40-200-CV
Protein and serum free medium, PF medium, Corning®
Milieux de culture cellulaire
PF medium is optimised for the cultivation of Chinese Hamster Ovary (CHO) cells, along with many derivatives of this parent line PF medium is a proprietary serum-free and protein-free growth medium that does not contain any hormones or growth factors. It medium supports superior growth and viability over long term passages in both adherent and suspension culture, and is formulated without L-glutamine.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité