EDTA, 0.5M pH 8.0, Corning®
Fournisseur: Corning
46-034-CI
MDTC46-034-CIEA 55.95 EUR InStock
MDTC46-034-CI
EDTA, 0.5M pH 8.0, Corning®
Tampons Tampons de culture de tissus
EDTA (ethylenediaminetetraacetic acid) is a polyamino carboxylic acid. It is a colorless, water-soluble solid and is useful as a chelating agent due to its ability to "sequester" metal ions such as Ca2+ and Fe3+.  These cations are responsible for various undesirable effects in molecular biological and cell culture processes. After being bound by EDTA, these metal ions remain in the solution but exhibit diminished reactivity.  The use of EDTA therefore allows for greater control over many experiments.
  • Complete Certificate of Analysis available for each production lot, along with full formulation information
  • Produced under the highest industry standards to ensre superior results.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité