Spectroquant® Prove Plus VIS and UV/VIS Spectrophotometers
Fournisseur: Merck
Spectroquant®
1.73026.0001EA 4774 EUR InStock
1.73026.0001 1.73028.0001 1.73027.0001
Spectroquant® Prove Plus VIS and UV/VIS Spectrophotometers
Spectrophotomètres et Photomètres Spectrophotomètres UV-Visible
The Spectroquant Prove 100/300/600 plus provides high quality and great value for money for your daily analyses.
The Spectroquant Prove plus family is our series of cutting-edge spectrophotometers for easy analysis of water, food and beverage, and environmental samples. All Spectroquant Prove plus instruments are pre-programmed for over 200 test Kits and automatic bar code recognition ensures the correct method parameters are applied and all essential information such as lot number, expiry date, and automatic calibration updates are recorded, making the use of analytical test Kits as easy as never before.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité