Synonyms:
Alcool éthylique
1.00986.1000EA
124
EUR
InStock
1.00986.1000
1.00986.9950_T
MERP1.00986.2500_T
1.00986.6025
MERP1.00986.6025_T
MERP1.00986.9180_T
MERP1.00986.6010_T
1.00986.2500
1.00986.9022
MERP1.00986.1000_T
1.00986.9950
1.00986.6010
MERP1.00986.1000DP
1.00986.6010DP
1.00986.6025DP
MERP1.00986.2500DP
1.00986.9180
Éthanol absolu, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP pouvant être utilisé comme excipient, SAFC®
Éthanol
Other residual solvents (ICH Q3C) excluded by production process.
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities).
Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Certifications: Conforms to Ph Eur, BP, JP, USP