1.20083.0500EA
0
EUR
InStock
1.20083.0500
1.20083.0100_T
1.20083.0500_T
1.20083.0100
Eshmuno® CPX, résine pour la séparation de racémates, pour la chromatographie
Eshmuno® CPX
Eshmuno® CPX media is a strong cation exchanger built on the proven Eshmuno® resin technology.
- Excellent mAb monomer/aggregate separation efficiency
- High resolution intermediate purification
- Superior dynamic binding capacity
- High throughput purification
- Rigid base bead for easy packing
- Outstanding pressure flow behavior