Titrator, Karl Fischer, coulometric, C30
Fournisseur: Mettler - Toledo
51105515
METN51105515EA 0 EUR InStock
METN51105515
Titrator, Karl Fischer, coulometric, C30
Titrateurs
This all-round Karl Fischer coulometer is a flexible and powerful instrument for fast and precise water determination of samples from any industry with a water content between 1 ppm and 5%. Up to 14 samples can be titrated in unattended operation with the Stromboli oven sample changer. Repeatability and efficiency is significantly improved and operation becomes simple, as Stromboli is fully controlled by the titrator.
  • Reagent Control monitors three different parameters and replaces the reagent when necessary in One Click
  • Flexible methods (8 pre-defined, max. 120 user methods) allow multiple blank and sample series with statistics
  • Color Touchscreen with One Click™ user interface
  • User-specific home screens with Shortcuts for One Click™ start of any task
  • Coulometric bromine index determination
  • 200 ml titration vessel
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité