Super Erecta® Industrial Wire Shelf, Metroseal Gray Epoxy
Fournisseur: Metro International
1848NK4
METO1848NK4EA 0 EUR InStock
METO1848NK4
Super Erecta® Industrial Wire Shelf, Metroseal Gray Epoxy
Mobilier Étagères
Neutral aesthetic for the lab environment, now with corrosion and antimicrobial protection.
  • Shelves can be repositioned at precise 1" (25 mm) increments along the length of the numbered posts
  • Open-wire design minimizes dust accumulation, allows a free circulation of air, and greater visibility of stored items
Corrosion resistant Metroseal Gray epoxy finish features Microban® antimicrobial product protection to keep the product 'cleaner between cleanings'.
12-year warranty against rust and corrosion.
Informations de commande: Posts and accessories sold separately.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité