Multi-parameter meters, bench, SevenExcellence™ S479 / S975-uMix
Fournisseur: Mettler - Toledo
30092988 30092989
METT30092988EA 6840 EUR InStock
METT30092988 METT30092989
Multi-parameter meters, bench, SevenExcellence™ S479 / S975-uMix
Electrochemistry Meters pH-mètres et multi-paramètres
Modular triple-channel instruments with touch screen operation, which can easily handle the most complex applications and meet stringent requirements in regulated markets.
  • Allows for measurements of 1, 2 or 3 parameters simultaneously, either in the same or different samples
  • Extra security thanks to sophisticated user management and Intelligent Sensor Management (ISM®)
  • Integrated interfaces (USB, RS232 and Ethernet) for data exchange
  • uPlace™ electrode holder (included) can be operated with one hand
  • Large 7 inch colour display and intuitive menu guidance in 10 languages
  • EasyDirect™ pH PC software for transferring real time measurement data and archiving the results
  • GLP support functions: PIN protection, GLP printout format with all GLP relevant information and monitoring of measurement limits
S479 allows users to measure pH/mV, rel. mV/ORP, conductivity (TDS, salinity, resisitivity, conductivity ash) as well as dissolved oxygen (using digital or polarographic technology).
S975 allows users to measure pH/mV, rel. mV/ORP, conductivity (TDS, salinity, resisitivity, conductivity ash), dissolved oxygen (using digital or polarographic technology), and the concentration and activity of ions in solution.
SevenExcellence meters fit three modules (expansion units) in any composition and order. They offer the highest possible flexibility as users can expand the instrument at any time, adding new measurement parameters.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité