CPC® Quick-Disconnect Fittings, Sanitary Bodies and Inserts
Fournisseur: CPC (Colder Products Company)
MFLX31604-27EA 41.3 EUR InStock
MFLX31604-27 MFLX31604-26 MFLX00296-OOBLK MFLX00296-OPBLK MFLX31604-28 MFLX00296-OQBLK MFLX31604-30 MFLX31604-34 MFLX31604-26BLK MFLX31604-32 MFLX31604-28BLK MFLX31604-27BLK MFLX00296-OQ
CPC® Quick-Disconnect Fittings, Sanitary Bodies and Inserts
Raccords et connecteurs
Attach directly to your sanitary hardware assemblies!
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • USP Class IV materials
  • Fully sterilisable
These couplings allow you to integrate disposables with your existing stainless steel processing equipment. Both couplings and inserts are available with standard sanitary terminations to attach to your system with a sanitary gasket and tri-clover clamps. All models feature USP Class VI platinum-cured silicone gaskets.
These quick-disconnect couplings require a large fitting body and large insert (order separately).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité