Masterflex® High-Purity Filter Cartridge Housings, Polypropylene, Avantor®
Fournisseur: Avantor Fluid Handling
01508-24
MFLX01508-24EA 130 EUR InStock
MFLX01508-24
Masterflex® High-Purity Filter Cartridge Housings, Polypropylene, Avantor®
Filtres Cartouches filtrantes
Filter housing is ideal for in-line process filtration systems.
  • Constructed of durable polypropylene (PP)
This #12 housing fits submicronic cartridges with 222 O-ring sealing caps. The pure PP housing contains no filters, lubricants, or plasticizers. Housing features an O-ring sealed PP drain plug in the sump with a second vent for differential pressure.
Maximum temperature is 100 °F (38 °C); maximum pressure is 100 psi. Inlet/outlet ports are 3/4" NPT(F); length is 14-3/8". Includes VITON O-ring.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité