Masterflex® Double-Open-End Granular Activated Carbon Filter Cartridges, Avantor®
Fournisseur: Avantor Fluid Handling
01508-92
MFLX01508-92EA 26.4 EUR InStock
MFLX01508-92
Masterflex® Double-Open-End Granular Activated Carbon Filter Cartridges, Avantor®
Filtres Cartouches filtrantes
Remove odors and chlorine taste from water.
  • Constructed with granular-activated carbon
Granular Activated Carbon Cartridges remove odors and chlorine taste from water. Approximately 10-oz of granular carbon is packed in the plastic tube. End caps have filters to protect carbon. Use at 1 GPM or lower. For cold water only. May initially contain leachable carbon dust; flush system well after installation. Maximum temperature is 125 °F (52 °C); maximum differential pressure is 70 psid**.
** For optional performance, change cartridges at 25 psid.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité