Masterflex® Variable-Area Flowmeter Valve Cartridges, Avantor®
Fournisseur: Avantor Fluid Handling
03218-70
MFLX03217-82EA 76.3 EUR InStock
MFLX03217-82 MFLX03217-84 MFLX03217-86 MFLX03217-88 MFLX03217-92 MFLX03217-96 MFLX03218-61 MFLX03218-62 MFLX03218-63 MFLX03218-64 MFLX03218-67 MFLX03218-68 MFLX03218-69 MFLX03218-72 MFLX03218-74 MFLX03218-70
Masterflex® Variable-Area Flowmeter Valve Cartridges, Avantor®
Débitmètres
Control your flow with the addition of a valve.
  • Aluminum is economical and good for general use with noncorrosive gases and liquids
  • 316 Stainless steel (SS) withstands higher temperatures and pressures, and features excellent chemical compatibility
Select a valve-cartridge that matches your frame. Insert these needle valves into the flowmeter frames to control flow rate. Choose a 16-turn, high-resolution metering valve-cartridge for greater precision of flow rate. If your flowtube is in units other than ml/min, convert them to ml/min and choose a valve within that flow range from the table below. See website and virtual catalog page for more details.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité