Accessories for Bürkle Stainless Steel Barrel Pumps for Flammable Liquids
Fournisseur: Avantor Fluid Handling
06043-80
MFLX06043-80EA 141 EUR InStock
MFLX06043-80
Accessories for Bürkle Stainless Steel Barrel Pumps for Flammable Liquids
Pompe Pompes pour conteneur/transfert
Compatible with the Bürkle Stainless Steel Barrel Pumps For flammable Liquids.
  • Safely transfer flammable and combustible liquids with ease
  • Constructed of conductive 304 Stainless Steel for enhanced durability and safety
Safely transfer flammable liquids with the stainless steel barrel pumps.
There are two pump models to choose from. The stainless steel barrel pumps with discharge tube feature a spout-type outlet. The drum pumps with discharge tube and stopcock are suitable for fluid transfer in which the outlet container is further from the filling container. The stopcock also allows for on/off fluid control. The removable hose is 1.2 m in length with a NW 8 mm ball valve and 12.8 mm diameter discharge nozzle.
The replacement 1.2 m stainless steel discharge hose stopcock is available as part number 06043-81 and the stainless steel discharge tube with spout-type outlet is available as part number 06043-80.
The hand-operated drum pumps are sold separately.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité