Bürkle Foot-Operated Barrel Pumps for Solvents
Fournisseur: Avantor Fluid Handling
06043-94
MFLX06043-94EA 1260 EUR InStock
MFLX06043-94
Bürkle Foot-Operated Barrel Pumps for Solvents
Pompe Pompes pour conteneur/transfert
Safely transfer corrosive solvents.
  • Stainless steel and PTFE construction withstands long-term exposure to aggressive solvents
  • Foot-operated bellows leaves both hands free for more stable container filling
  • Telescopic immersion tube adapts itself to a variety of container heights
  • Fits a 2" thread creating a closed system to contain harmful vapors
Use this foot-operated bellows pump to safely dispense solvents from drums up to 55 gal. (220 L). Choose between a pump with a spout or a discharge hose; both models include a stopcock to contain vapors and prevent dripping. Order optional antistatic grounding set for pumping flammable liquids.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité