Bürkle Compact Fluid Sampling Kit for Barrels, Canisters, and Tanks Accessories
Fournisseur: Avantor Fluid Handling
06044-36
MFLX06044-35EA 97.7 EUR InStock
MFLX06044-35 MFLX06044-36
Bürkle Compact Fluid Sampling Kit for Barrels, Canisters, and Tanks Accessories
Pompe Pompes pour conteneur/transfert
Accessories for the Compact Fluid Sampling Kits.
  • Eliminate cross-contamination while taking samples
  • Durable components suitable for use with viscous oils
  • Quick and comfortable sampling with hand-operated vacuum-piston pump
The compact fluid sampling systems greatly reduce the possibility of cross-contamination as the liquid only comes into contact with the suction hose and sample bottle. After the sample is taken, the bottles are sealed, and the suction tubing can quickly and easily be replaced. The small-diameter 8-mm hose can reach narrow, hard-to-access sampling areas. Both models can be used with the PTFE adapter for glass thread GL45 for enhanced chemical resistance and use with other sampling bottles.
Kit 06040-94 is for general sampling from barrels, canisters, tanks, and more, as the wetted materials are polyethylene. This kit can be with the 06040-27 HDPE sampling bottle and polyethylene suction hose 06044-36 (10 m roll) or 06044-37 (100 m roll).
Kit 06040-95 is for ultrapure sampling and ideal when maximum purity or chemical resistance is required. This pump can be used with the PTFE suction hose 06044-35 (5 m roll).
Sampling kits sold separately.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité