Masterflex® Fittings, Compression Nut Assembly with Ferrule, Avantor®
Fournisseur: Avantor Fluid Handling
06380-11
MFLX06380-11EA 30.5 EUR InStock
MFLX06380-11
Masterflex® Fittings, Compression Nut Assembly with Ferrule, Avantor®
Raccords et connecteurs
Can handle pressures up to 225 psi.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rated to 220 psi maximum at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité