Masterflex® Union Fittings, Compression, Tee, Avantor®
Fournisseur: Avantor Fluid Handling
06382-30
MFLX06382-30EA 58.1 EUR InStock
MFLX06382-30
Masterflex® Union Fittings, Compression, Tee, Avantor®
Raccords et connecteurs
Excellent choice to connect rigid tubing systems.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Compression fittings offer a more permanent installation. Choose the size and material to suit your application needs.
Good chemical resistance. Usable from -50 to 250 °F (-46 to 121 °C).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité