Masterflex® Transfer Tubing, Tygon® Tygon® S3™ E-3603, Avantor®
Fournisseur: Avantor Fluid Handling
06407-99
MFLX06407-70EA 42.2 EUR InStock
MFLX06407-70 MFLX06407-71 MFLX06407-72 MFLX06407-73 MFLX06407-74 MFLX06407-75 MFLX06407-76 MFLX06407-77 MFLX06407-78 MFLX06407-79 MFLX06407-80 MFLX06407-81 MFLX06407-82 MFLX06407-83 MFLX06407-84 MFLX06407-85 MFLX06407-86 MFLX06407-87 MFLX06407-88 MFLX06407-89 MFLX06407-90 MFLX06407-91 MFLX06407-92 MFLX06407-93 MFLX06407-94 MFLX06407-95 MFLX06407-96 MFLX06407-98 MFLX06407-99
Masterflex® Transfer Tubing, Tygon® Tygon® S3™ E-3603, Avantor®
Tuyaux Transfer Tubing
Pliable formulation for sharp radius bending.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
This tubing is ideal for general laboratory use. It offers long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, and is nontoxic.
FDA compliant and made according to GMP.
Temperature range is -51 to 165 °F (-46 to 73 °C)
Can be sterilized with ethylene oxide or autoclave
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité