Masterflex® L/S® Precision Pump Tubing, C-Flex®, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX06424-13EA 68.9 EUR InStock
MFLX06424-13 MFLX06424-14 MFLX06424-16 MFLX06424-17 MFLX06424-18 MFLX06424-25
Masterflex® L/S® Precision Pump Tubing, C-Flex®, Avantor®
Tuyaux Tubes pour pompe péristaltique
Combines biocompatibility of silicone with chemical resistance similar to Tygon®.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité