Plastic Hand-Operated Threaded Hand Pump
Fournisseur: Avantor Fluid Handling
06433-60
MFLX06433-60EA 84.7 EUR InStock
MFLX06433-60
Plastic Hand-Operated Threaded Hand Pump
Pompe Pompes pour conteneur/transfert
Ideal for most acid/base or ionic salt applications.
  • Pump detergents, water, dilute alkalies, and waxes from drums or pails — 3/4" NPT(M) intake threads permit semi-permanent attachment
This threaded plastic hand-operated drum pump is ideal for pumping detergents, water, dilute alkalis, and waxes from drum pails. Constructed with a PVC body and hose, Hypalon® valves and seals, and LDPE tube, this pump offers 26 strokes/gallon (7 strokes/liter). Optional adapters allow secure attachment to a variety of containers. Order adapters 06433-62, -64, -66, and -68 separately.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité