Masterflex® Ismatec® Pump Tubing, Peroxide-Cured Silicone, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX06516-34EA 170 EUR InStock
MFLX06516-34
Masterflex® Ismatec® Pump Tubing, Peroxide-Cured Silicone, Avantor®
Tuyaux Tubes pour pompe péristaltique
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Silicone tubing blended with organic peroxide for biological applications. This economical tubing has long life, making it ideal for lab and general applications.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité