Masterflex® MasterSense™ MP1 PendoTECH™ Pressure Sensor Adaptor Kits, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX07526-90EA 1510 EUR InStock
MFLX07526-90 MFLX07526-91 MFLX07526-92
Masterflex® MasterSense™ MP1 PendoTECH™ Pressure Sensor Adaptor Kits, Avantor®
Pompe Pompes péristaltiques
Designed for simplified integration of a single PendoTECH™ pressure sensor into Masterflex® MasterSense™ pump systems.
  • Add critical pressure monitoring to your workflow
  • Specifically designed for PendoTECH™ single-use pressure sensors
  • Perfect for tangential-flow filtration, depth filtration, harvest, bulk transfer, chromatography and fill applications
  • IP 66 washdown housing
  • CE compliance
  • ABS housing for wipe-down and cleanability
asterSense™ M1 adaptors allow for simplified connection of PendoTECH™ single-use pressure sensors into MasterSense™ peristaltic pump systems. MP1 is designed for benchtop. Adaptor kits include everything needed for initial process.
Install sensors downstream of pump to detect static and dynamic process pressure and to avoid downtime.
Choose the started kit with the appropriate tubing size/process connection for your application, or purchase the MP1 unit and sensors ala cart.
Certifications: CE certified.
Informations de commande: PendoTECH™ pressure sensors allow for easy incorporation into single-use systems aseptic control. Order additional pressure sensors separately.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité