Masterflex® L/S® Variable-Speed Air-Powered Drive, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX07569-00EA 2110 EUR InStock
MFLX07569-00
Masterflex® L/S® Variable-Speed Air-Powered Drive, Avantor®
Pompe Pump Drives Peristaltic Pump Drives
Use where electrical power is unsafe or unavailable.
  • Flow range 3.6 to 3400 ml/min—low rate depends on drive rpm, tubing head, and tubing size
  • Easily vary speed by adjusting air pressure with regulator
  • Rugged 1/3-hp air motor requires little maintenance and generates less heat than an electric motor
  • IP44 rated—painted steel housing protects from gentle spray
Use this variable-speed air-powered drive whenever electrical power is unsafe or impractical. Careful grounding to protect from static electricity is vital for safe operation. Drive accepts nine pump head types: Standard, Easy-Load®, Easy-Load® II, Easy-Load® 3, Multichannel, High-Performance, Cartridge, PTFE-Tubing, and PTFE-Diaphragm.
Information de livraison: Includes air/speed regulator with 5-μm air filter, automatic lubricator, 0 to 30 psi pressure gauge, and muffler.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité