Pressure Sensor Transmitters
Fournisseur: Avantor Fluid Handling
MFLX19406-67EA 1630 EUR InStock
MFLX19406-67
Pressure Sensor Transmitters
Électrodes Électrodes, autres
The enhanced PendoTECH® pressure sensor transmitter connects seamlessly to the PendoTECH pressure sensor, delivering a 4 to 20 mA signal linear with pressure. It maintains its convenient DIN rail mounting design for easy installation and integration into bioprocess systems.
  • The PTR features an alarm output function with a dry contact relay output
  • The PTR supports the digital communication protocol of modbus TCP
  • It has a convenient DIN rail mounting design for easy integration
Two model options are available to deliver optimal performance based on pressure range. For lowest-pressure applications, the PTR 7,5 psi (MFLX19406-66) has a default 4 to 20 mA range of –11,5 to +7,5 psi. For higher pressure applications, the PTR 75 psi (MFLX19406-67) supports the full pressure range –11,5 to +75 psi.
Informations de commande: Selecting the narrowest range that meets the application requirements achieves better accuracy and repeatability. See data sheet for additional details.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité