Masterflex® Single-Use Tubing Pinch Clamp, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX19999-99EA 47.2 EUR InStock
MFLX19999-99
Masterflex® Single-Use Tubing Pinch Clamp, Avantor®
Pinces Clips pour tubes
Swing-away lockdown nut provides ease of use.
  • Higher tensile strength than polysulfone—less likely to break
  • Offers quick release from locked position
  • Ideal for biopharma applications that require gamma irradiation or autoclave cycles
This clamp accepts tubing from 0.187 to 0.500" inner diameters. The recommended wall thickness is 0.085 to 0.200". Tubing materials that can be used include silicone, TPE TPV, PVC, and other rubbers. The maximum hardness for correct functioning of the clamp is 70 Shore A. May be installed on existing tube assembliesMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité