Masterflex® Strainer Mesh for Low-Cost In-Line Strainer Systems, Avantor®
Fournisseur: Avantor Fluid Handling
29595-31 29595-33 29595-35 29595-39 29595-41 29595-43 29595-45 29595-47 29595-49 29595-51 29595-55 29595-57
MFLX29595-31EA 18.7 EUR InStock
MFLX29595-31 MFLX29595-33 MFLX29595-35 MFLX29595-39 MFLX29595-41 MFLX29595-43 MFLX29595-45 MFLX29595-47 MFLX29595-49 MFLX29595-51 MFLX29595-55 MFLX29595-57
Masterflex® Strainer Mesh for Low-Cost In-Line Strainer Systems, Avantor®
Filtres Strainers
Easily customize strainer system to suit your application.
  • Choose multiple mesh and micron sizes options based on required filtration area
  • Constructed of 304 stainless steel
Build your own economical filtration system to filter out particulates from your fluids. Available in three classes: low-profile, intermediate, and large. Modify or change components of the same class to meet the demands of your application.
Low-Profile Strainers are ideal for small-scale low-volume filtration. Use in research applications, cleaning equipment, and laboratoryMore Product Information
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité