Carbon-Impregnated Pleated Polyester Cartridges for "Big Blue" Housing
Fournisseur: Avantor Fluid Handling
29830-33
MFLX29830-33EA 87.3 EUR InStock
MFLX29830-33
Carbon-Impregnated Pleated Polyester Cartridges for "Big Blue" Housing
Filtres Cartouches filtrantes
Pleated filter gives you more surface area for higher flowrate and longer life.
  • Constructed of Carbon-Impregnated Pleated Polyester
The pleated design offers higher flow rates and dirt-loading capacities than shorter-life block cartridges. The non-cellulose media resists chemical attack and biofilm build-up making the filter ideal for both chlorinated and non-chlorinated water treatment applications. Common applications include post reverse osmosis polishing and well water treatment.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité