PrimeLock® High-Purity PFA Fittings, Standard Tees
Fournisseur: Avantor Fluid Handling
30492-81
MFLX30492-81EA 172 EUR InStock
MFLX30492-81
PrimeLock® High-Purity PFA Fittings, Standard Tees
Raccords et connecteurs
Ultra-clean and reliable fittings for highly critical and corrosive chemicals.
  • Broad chemical compatibility with PFA wetted components
  • Three nonwetted backup seals prevent leaks without introducing sources of entrapment or flow interruptions
  • Audible and visual checks guarantee proper connections
  • Heat not required for fitting installation
PFA wetted fittings handle critical chemical applications for manufacturers and laboratories. Tubing connections offer a primary seal and multiple safety backups with nonwetted inserts, reducing entrapment, leak points, and flow interruptions in the lines. Installation tools offer consistent and repeatable assembly without using heat to produce a flare in the tubing. If necessary, fittings can be installed without insertion tools by using a hot air gun. Installation tools and accessories are designed for a proper joint fit and superior connection integrity for all PrimeLock fittings.
Information de livraison: Fittings include one set of nuts, inserts, and connection verification rings.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité