Masterflex® Adapter Fittings, Hose Barb to Sanitary Clamp, Straight, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX30700-62EA 74.4 EUR InStock
MFLX30700-62
Masterflex® Adapter Fittings, Hose Barb to Sanitary Clamp, Straight, Avantor®
Raccords et connecteurs
Meets FDA requirements and 3A sanitary standards.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Easily connects pipes or fittings of different types
  • Compatible with standard 304L fitting clamps
  • Ideal for pharmaceutical industry
These barbed fitting offers tight seal on hoses when used with a hose clamp (not included). Ideal for pharmaceutical industry, and chemical compatibility for use with corrosive acids and bases.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité