Masterflex® Adapter Fittings, Sanitary Clamp to Female Threaded, Straight, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX31200-13EA 169 EUR InStock
MFLX31200-13
Masterflex® Adapter Fittings, Sanitary Clamp to Female Threaded, Straight, Avantor®
Raccords et connecteurs
Maintain sanitary conditions.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp® connections
  • Excellent chemical resistance; low leachable characteristics
These fittings are ideal for use with the Masterflex L/S tubing pumps.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité