CPC® High-Flow Quick-Disconnect Fittings, Elbow Hose Barb Inserts
Fournisseur: Avantor Fluid Handling
31308-75BLK
MFLX31303-51EA 20.2 EUR InStock
MFLX31303-51 MFLX31303-55 MFLX31303-56 MFLX31307-56 MFLX31303-51BLK MFLX31303-55BLK MFLX31303-56BLK MFLX31307-56BLK MFLX31308-75BLK
CPC® High-Flow Quick-Disconnect Fittings, Elbow Hose Barb Inserts
Raccords et connecteurs
No more leaks - valved couplings automatically shut off flow.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many ¹/₂" flow couplings in a ³/₈" body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 stainless steel valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 stainless steel latch springs and EPDM seals.
For a complete coupling, order one fitting body and one insert.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité