Adapter Fittings, Compression to Male Threaded, Elbows
Fournisseur: Avantor Fluid Handling
31412-86
MFLX31412-86EA 19.7 EUR InStock
MFLX31412-86
Adapter Fittings, Compression to Male Threaded, Elbows
Raccords et connecteurs
Maintain a permanent connection with these adapters.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These threaded fittings are suitable for permanent installation and can used with your rigid tubing for high-pressure applications.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité