Masterflex® ADCF Polypropylene Sanitary Adapters, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX31513-18EA 78.3 EUR InStock
MFLX31513-18
Masterflex® ADCF Polypropylene Sanitary Adapters, Avantor®
Raccords et connecteurs
These adapters are perfect to transition from sanitary clamp connections to a barb for flexible tubing.
  • Constructed of durable Polypropylene
All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemical resistance and may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI. Available with 3/8" or 1/2" hose barbs and 3/4" mini or 1-1/2" standard sanitary clamp connections.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité