Venture Tech PFA Sanitary Clamps, 90° Elbow
Fournisseur: Avantor Fluid Handling
31530-02
MFLX31530-02EA 171 EUR InStock
MFLX31530-02
Venture Tech PFA Sanitary Clamps, 90° Elbow
Raccords et connecteurs
Ideal for extremely corrosive materials—PFA will not rouge or pit.
  • Inert, nonstick surfaces—perfect for fragile and metal-sensitive proteins
  • Employ regular steam-in-place cycles without worry
  • Fully compatible with other Tri-Clamp® connections
  • Protective overmolding reinforces clamp connection sites
  • Full static vacuum capability at 300 °F (149 °C)
Sanitary applications that use highly corrosive chemicals require fittings that can endure the punishment. These food and pharmaceutical quality PFA fittings are easily cleaned so you will minimize downtime, cross contamination, and yield loss. Autoclavable.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité