GF Signet® Paddle-Wheel Flow System Installation Fittings
Fournisseur: Avantor Fluid Handling
32511-80
MFLX32511-80EA 355 EUR InStock
MFLX32511-80
GF Signet® Paddle-Wheel Flow System Installation Fittings
Débitmètres
Build a complete paddle-wheel flow sensor system customizable to your application.
  • Modular system for a customizable approach to flow measurement
  • Ideal for easy installation
The GF Signet paddle-wheel flow system is comprised of three components. Select the flow sensor, installation fitting, and flow meter that suit your application needs for a customizable approach to flow measurement.
Step One: Choose a Flow Sensor
Step Two: Choose an Installation Fitting
Step Three: Choose a Flow Meter
Flow sensor, installation fitting, and flow meter sold separately.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité