Masterflex® Flow Indicators and Switches for Liquids, Avantor®
Fournisseur: Avantor Fluid Handling
32880-20
MFLX32880-20EA 610 EUR InStock
MFLX32880-20
Masterflex® Flow Indicators and Switches for Liquids, Avantor®
Flow Indicators
Easily view flow through clear plastic lens.
  • Constructed with polypropylene (PP) housing
Flow indicators feature a paddle-wheel that spins at a rate proportional to the flow. Lens can be removed with a 1/4 turn for easy cleaning. Use with fluids that have a viscosity less than 34 centipoise (150 µm filter is r ecommended for protection).
Indicators feature a bright orange rotor that spins to provide visual verification.
Indicators/Sensors feature a Hall-effect sensor that produces a 4.5 to 24 VDC pulse output. Maximum current source output is 70 mA, and frequency output range is approximately 25 Hz at low flow to 225 Hz at high flow.
Indicators/Switches offer extra level of reliability and protection with a built-in switch. Each Single Pole Double Throw (SPDT) switch is rated for 1 amp, 24 VDC resistive. Repeatability is ±2% ; accuracy is ±5%. An LED lights when the set point is reached for easy field calibration.
Information de livraison: Includes 24" (61 cm) PVC jacketed cable.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité