John Guest® Push-to-Connect Check Valves, Acetal
Fournisseur: Avantor Fluid Handling
MFLX34006-64EA 235 EUR InStock
MFLX34006-64
John Guest® Push-to-Connect Check Valves, Acetal
Raccords et connecteurs
Get a secure connections with quick disconnection.
  • Constructed of durable acetal
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité