John Guest polypropylene push-to-connect fittings
Fournisseur: Avantor Fluid Handling
34007-52
MFLX34007-48EA 46.2 EUR InStock
MFLX34007-48 MFLX34007-49 MFLX34007-51 MFLX34007-52
John Guest polypropylene push-to-connect fittings
Raccords et connecteurs
Quick, leakproof connection.
  • Polypropylene and EPDM materials meet NSF 51, and NSF 61 requirements for food-grade applications, and is FDA compliant
Push-to-connect fittings offer secure connections and quick disconnection. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semi-rigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing.
Adapters (sold separately) let you permanently modify basic fittings for customized installations.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité