Masterflex® Luer Lock Fittings, Stopcock, Avantor®
Fournisseur: Avantor Fluid Handling
40619-99
MFLX31200-70EA 64.5 EUR InStock
MFLX31200-70 MFLX31200-90 MFLX31200-91 MFLX31200-92 MFLX40619-99
Masterflex® Luer Lock Fittings, Stopcock, Avantor®
Vannes Ball Valves
Build leakproof fluid handling systems.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
These one-way stopcocks feature an all-black PVDF construction.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité